missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, sodium salt trihydrate, 99+%, for biochemistry

Product Code. 12103761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12103761 250 g Plastic bottle 250g
12183751 1 kg Plastic bottle 1kg
12193751 5 kg Plastic bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12103761 Supplier Thermo Scientific Acros Supplier No. 446792500
  • ONLINE EXCLUSIVE PRICE
    39.25 €
    38.10 € / 250g
    Save 1.15 €
    2% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6131-90-4 | C2H3NaO2·3H2O | 136.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H3NaO2·3H2O
Molecular Weight (g/mol) 136.08
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Absorbance 0.05 max. (3M) at 260nm, 0.05 max. (3M) at 280nm
Melting Point 58.0°C
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula CH3COONa·3H2O
Quantity 250 g
Merck Index 15,8709
Solubility Information Solubility in water: 762g/L (20°C). Other solubilities: 1g/19mL alcohol
Formula Weight 136.08
Percent Purity 99+%
Grade Biochemistry
Insoluble Matter 0.005% max. (in water)
Chemical Name or Material Acetic acid, sodium salt trihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.