missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, sodium salt trihydrate, for HPLC, Thermo Scientific Chemicals

Product Code. 11904911 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11904911 250 g 250g
11994901 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11904911 Supplier Thermo Scientific Supplier No. 446202500
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6131-90-4, C2H9NaO5, 136.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.08
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
IUPAC Name sodium acetate trihydrate
SMILES O.O.O.[Na+].CC([O-])=O

Specifications

Melting Point 58.0°C
Quantity 250 g
Linear Formula CH3COONa·3 H2O
Green Features ACT Ecolabel (Gold)
Merck Index 15, 8709
Solubility Information Solubility aq. soln.: 762 g/L (20°C). Other solubilities: 1 g/19 mL alcohol
Formula Weight 136.08
Grade HPLC
Chemical Name or Material Acetic acid, sodium salt trihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.