missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid solution, 0.1M, Honeywell™

Product Code. 15624130 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15624130 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15624130 Supplier Honeywell-Fluka Supplier No. 342541L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric, 0.1 M CH3COOH (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 64-19-7
Molecular Formula C2H4O2
Linear Formula CH3CO2H
MDL Number MFCD00036152
UN Number NONH for all modes of transport
Beilstein 506007
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(O)=O
Concentration 0.1M
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.05
PubChem CID 176
ChEBI CHEBI:15366
Formula Weight 60.05g/mol
Quantity 1 L
Chemical Name or Material Acetic acid solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.