missing translation for 'onlineSavingsMsg'
Learn More

Acetic anhydride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99% (GC), Solstice

Product Code. 15643940 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Unit Size:
Pack of 4
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15643940 4 x 2.5 L Pack of 4
15663940 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 15643940 Supplier Honeywell-Fluka Supplier No. 332144X2.5L
  • ONLINE EXCLUSIVE PRICE
    461.00 €
    433.02 € / Pack of 4
    Save 27.98 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

ACS Reagent, with ≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00008705
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
PubChem CID 7918
ChEBI CHEBI:36610
IUPAC Name acetyl acetate
SMILES CC(=O)OC(=O)C

Specifications

Density 1.08g/mL
Flash Point 49°C (120.2°F)
Linear Formula (CH3CO)2O
Quantity 4 x 2.5 L
UN Number UN1715
Formula Weight 102.09
Percent Purity ≥99% (GC)
Chemical Name or Material Acetic anhydride
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acetic anhydride
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302+H332-Harmful if swallowed or if inhaled
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.