missing translation for 'onlineSavingsMsg'
Learn More

Acetic-d3 acid-d, for NMR, 99.5 atom % D

Product Code. 10511374 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
50 mL
Unit Size:
25mL
50mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10511374 50 mL 50mL
10605802 5 mL 5mL
10762971 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10511374 Supplier Thermo Scientific Acros Supplier No. 166210500
  • ONLINE EXCLUSIVE PRICE
    327.00 €
    317.00 € / 50mL
    Save 10.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1186-52-3 | C2H4O2 | 64.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1186-52-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 64.08
MDL Number MFCD00051051
InChI Key QTBSBXVTEAMEQO-GUEYOVJQSA-N
Synonym acetic acid-d4, acetic-d3 acid-d, 2h3 acetic 2h acid, tetradeuteroacetic acid, 2 h? ethan 2 h oic acid, acetic acid d4, 2h4 acetic acid, cd3cood, acetic acid d, acetic acid-d4, 99.5 atom % d
PubChem CID 2723903
IUPAC Name deuterio 2,2,2-trideuterioacetate
SMILES [2H]OC(=O)C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Density 1.1300g/mL
Deuterium (D) 99.4% min.
Flash Point 40°C
Refractive Index 1.3709
Specific Gravity 1.13
Water 0.1% max. (Coulometric)
Quantity 50 mL
Melting Point 15.0°C to 16.0°C
Boiling Point 115.5°C
Linear Formula CD3CO2D
Beilstein 02, III, 203
Formula Weight 64.08
Grade NMR
Chemical Name or Material Acetic-d3 acid-d, 99.5 atom%D
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.