missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetamide, 97%

Product Code. 11915531 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11915531 500 g Glass bottle 500g
11398141 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11915531 Supplier Thermo Scientific Acros Supplier No. 442545000
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5977-14-0 | C4H7NO2 | 101.105 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5977-14-0
InChI Key GCPWJFKTWGFEHH-UHFFFAOYSA-N
Synonym acetoacetamide, acetylacetamide, acetoacetic acid amide, butanamide, 3-oxo, 3-oxobutanamid, unii-6o9u7cfy0t, 6o9u7cfy0t, 3-oxobutyramide, acmc-20alv9
PubChem CID 80077
ChEBI CHEBI:28515
IUPAC Name 3-oxobutanamide
SMILES CC(=O)CC(=O)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -53.0°C to +56.0°C
Color White to Yellow
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C4H7NO2
Linear Formula CH3C(O)CH2C(O)NH2
Quantity 500 g
Molecular Weight (g/mol) 101.105
Formula Weight 101.11
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Acetoacetamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.