missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetanilide, 99%

Product Code. 10764781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
1 kg
Unit Size:
1kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10764781 1 kg 1kg
10611063 5 g 5g
10234510 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10764781 Supplier Thermo Scientific Acros Supplier No. 102360010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 102-01-2 | C10H11NO2 | 177.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-01-2
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.2
MDL Number MFCD00008780
InChI Key DYRDKSSFIWVSNM-UHFFFAOYSA-N
Synonym acetoacetanilide, butanamide, 3-oxo-n-phenyl, acetoacetic anilide, acetoacetamidobenzene, acetylacetanilide, acetoacetylaniline, acetoacetanilid, n-phenylacetoacetamide, acetoacetic acid anilide, n-acetylacetyl aniline
PubChem CID 7592
IUPAC Name 3-oxo-N-phenylbutanamide
SMILES CC(=O)CC(=O)NC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 83.0°C to 86.0°C
Color White
Flash Point 162°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3COCH2CONHC6H5
Quantity 1 kg
Beilstein 12, 518
Merck Index 15, 57
Solubility Information Solubility in water: 5g/L (20 c). Other solubilities: soluble in acids and alkali hydroxide solutions,soluble in alcohol,chloroform,ether,hot benzene,and hot petroleum ether
Formula Weight 177.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Acetoacetanilide, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.