missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetyl coenzyme A sodium salt, >90%, MP Biomedicals™

Product Code. 11483031 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483031 50 mg 50mg
11463031 10 mg 10mg
11473031 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483031 Supplier MP Biomedicals Supplier No. 0215022450
  • ONLINE EXCLUSIVE PRICE
    1985.00 €
    1878.45 € / 50mg
    Save 106.55 €
    5% Off
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 102029-52-7
Molecular Formula C25H40N7NaO18P3S
Molecular Weight (g/mol) 874.597
InChI Key RSOOKMDOLPTBRH-UHFFFAOYSA-N
PubChem CID 133112705
IUPAC Name S-[2-[3-[[4-[[[5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] 3-oxobutanethioate;sodium
SMILES CC(=O)CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O.[Na]

Specifications

Color Yellow
Assay Percent Range >90%
Quantity 50 mg
Formula Weight 901.6 (free acid)
Physical Form Solid
Chemical Name or Material Acetoacetyl coenzyme A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.