missing translation for 'onlineSavingsMsg'
Learn More

Acetochlor, TRC

Product Code. 30314062
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314062 250 mg 250mg
30313215 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30314062 Supplier Toronto Research Chemicals Supplier No. TRCA162500250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetochlor
CAS 34256-82-1
Molecular Formula C14 H20 Cl N O2
Quantity 250 mg
Synonym o-Acetotoluidide, 2-chloro-N-(ethoxymethyl)-6'-ethyl- (8CI), 2-Chloro-N-(ethoxymethyl)-N-(2-ethyl-6-methylphenyl)acetamide, 2-Chloro-N-(ethoxymethyl)-6'-ethyl-o-acetotoluidide, 2-Methyl-6-ethyl-N-ethoxymethyl-2-chloroacetanilide, 2'-Ethyl-6'-methyl-N-(ethoxymethyl)-2-chloroacetanilide, Acenit, Acetal, Acetal (herbicide), Acetochlor, Acetor 50 EC, Azetochlor, Guardian, Guardian (herbicide), Harness, Henaisi, MG 02, MON 097, Nevirex, Relay, Relay (herbicide), Riley, Surpass, Trophee, Trophy, Warrant
InChI Formula InChI=1S/C14H20ClNO2/c1-4-12-8-6-7-11(3)14(12)16(10-18-5-2)13(17)9-15/h6-8H,4-5,9-10H2,1-3H3
SMILES CCOCN(C(=O)CCl)c1c(C)cccc1CC
IUPAC Name 2-chloro-N-(ethoxymethyl)-N-(2-ethyl-6-methylphenyl)acetamide
Molecular Weight (g/mol) 269.77
Formula Weight 269.1183
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.