missing translation for 'onlineSavingsMsg'
Learn More

Acetoin (~90%), TRC

Product Code. 30005391
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005391 50 g 50g
30011040 5 g 5g
30006026 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005391 Supplier Toronto Research Chemicals Supplier No. TRCA16377850G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-86-0
Formula Weight 88.0524
InChI Formula InChI=1S/C4H8O2/c1-3(5)4(2)6/h3,5H,1-2H3
IUPAC Name 3-hydroxybutan-2-one
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
SMILES CC(O)C(=O)C
Synonym 2-Butanone, 3-hydroxy-, 3-Hydroxy-2-butanone, (±)-3-Hydroxybutan-2-one, (±)-Acetoin, 1-Hydroxyethyl methyl ketone, 2,3-Butanolone, 2-Hydroxy-3-butanone, 3-Hydroxybutanone, 3-Hydroxyl-2-butanone, 3-Oxo-2-butanol, Acetoin, Acetoine, Acetyl methyl carbinol, DL-Acetoin, Dimethylketol, Methanol, acetylmethyl-, NSC 7609, γ-Hydroxy-β-oxobutane
Chemical Name or Material Acetoin

Specifications

Quantity 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.