missing translation for 'onlineSavingsMsg'
Learn More

Acetone, 99.6%, for spectroscopy ACS

Product Code. 10269830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10269830 500 mL Glass Bottle 500mL
10470831 1 L Glass Bottle 1 L
10045800 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10269830 Supplier Thermo Scientific Acros Supplier No. 400105000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67-64-1 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95.0°C
Absorbance 0.01 max. at 400nm, 0.02 max. at 350nm, 0.1 max. at 340nm, 1 max. at 330nm
CAS Min % 99.5
CAS Max % 100.0
Decomposition Information >4°C
Density 0.7900g/mL
Boiling Point 56.0°C
Quantity 500 mL
Flash Point −20°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3585
Assay Percent Range 99.6%
Residue after Evaporation 0.001% max.
Specific Gravity 0.79
Titratable Acid 0.0003 meq/g max.
Linear Formula CH3COCH3
Titratable Base 0.0006 meq/g max.
Vapor Pressure Vapor Pressure: 247mbar at 20°C
Viscosity 0.32 mPa.s (20°C)
Beilstein 01, 635
Water 0.5% max. (K.F.)
Fieser 02,13; 03,4; 06,9
Merck Index 15, 65
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
Formula Weight 58.08
Percent Purity 99.6%
Grade ACS
Chemical Name or Material Acetone, Reagent ACS, Spectro Grade
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.