missing translation for 'onlineSavingsMsg'
Learn More

Acetone a.r. For Laboratory Use, Chem Lab

Product Code. 15927996
Change view
Click to view available options
:
Acetone a.r. For Laboratory Use, 2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15927996 Acetone a.r. For Laboratory Use, 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15927996 Supplier Chem-Lab Supplier No. CL00.0114.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point -94°C
CAS 67-64-1
Color Colorless
Acidity < 0.0003 meq/g
Density 0.791 g/mL
Quantity 2.5 L
Flash Point -20 °C
Packaging Polyethylene
Physical Form Liquid
Residue after Evaporation < 0.0005%
Assay Percent Range > 99.5%
Molecular Formula C3H6O
MDL Number MFCD00008765
Water < 0.2%
Boiling Point 56°C
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
Grade Analytical
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone a.r.
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.