missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, BAKER ANALYZED, J.T. Baker™
109.25 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
2.5 L
Packaging:
HDPE
Description
- Exceeds the requirements set forth by the American Chemical Society (ACS)
- Label provides an actual Lot Analysis as a guarantee of purity and consistency
- Analytical methods used for these determinations are based on standard test procedures described in technical publications
- Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability
Specifications
pH | 5 to 6 (at 20°C) |
Melting Point | -94.9°C |
Absorbance | Max. 1.00 at 330nm, Max. 0.10 at 340nm, Max. 0.02 at 350nm, Max. 0.01 at 400nm |
CAS | 67-64-1 |
Color | Colorless |
Density | 0.79g/cm3 at 20°C |
Boiling Point | 55.8°C to 56.6°C |
Quantity | 2.5 L |
Explosion Limit | 2.6 - 13 Vol % |
Flash Point | −20°C |
Show More |
Safety and Handling
Product Identifier
- Acetone
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : acetone
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'