missing translation for 'onlineSavingsMsg'
Learn More

Acetone-d{6}, 99.9% (Isotopic), contains 1% v/v TMS

Product Code. 11324618
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324618 10 g 10g
11314618 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11324618 Supplier Thermo Scientific Alfa Aesar Supplier No. 042458.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 666-52-4 | C3H6O | 64.117 g/mol

Acetone-d{6} is used to produce Perdeuterio-aceton-cyanhydrin at temperature of 15 - 20°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 666-52-4
Molecular Formula C3H6O
Molecular Weight (g/mol) 64.117
MDL Number MFCD00044635
InChI Key CSCPPACGZOOCGX-WFGJKAKNSA-N
Synonym acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
PubChem CID 522220
ChEBI CHEBI:78217
IUPAC Name 1,1,1,3,3,3-hexadeuteriopropan-2-one
SMILES CC(=O)C

Specifications

Physical Form Liquid
Color Colorless
Density 0.872 g/mL
Flash Point −17°C (1°F)
Odor Acetone-like
Refractive Index 1.3554
Quantity 10 g
Melting Point -94°C
Boiling Point 56°C
UN Number UN1090
Solubility Information Fully miscible in water.
Formula Weight 64.13
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Contains 1% v/v TMS
Chemical Name or Material Acetone-d6, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.