missing translation for 'onlineSavingsMsg'
Learn More

Acetone-d{6}, 99.9% (Isotopic)

Product Code. 11317218
Change view
Click to view available options
Quantity:
10 g
5 x 10 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
11317218 10 g 10g
11327218 5 x 10 g 50g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11317218

Brand: Thermo Scientific Alfa Aesar 016797.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

NMR solvent | CAS: 666-52-4 | C3H6O | 64.117 g/mol

Acetone-d{6} is employed as an NMR solvent. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 666-52-4
Molecular Formula C3H6O
Molecular Weight (g/mol) 64.117
MDL Number MFCD00044635
InChI Key CSCPPACGZOOCGX-WFGJKAKNSA-N
Synonym acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
PubChem CID 522220
ChEBI CHEBI:78217
IUPAC Name 1,1,1,3,3,3-hexadeuteriopropan-2-one
SMILES CC(=O)C

Specifications

Physical Form Liquid
Color Colorless
Density 0.872 g/mL
Flash Point -17°C (1°F)
Odor Acetone-like
Refractive Index 1.3554
Quantity 10 g
Melting Point -94°C
Boiling Point 56°C
UN Number UN1090
Solubility Information Soluble in water.
Formula Weight 64.13
Percent Purity 99.9%
Chemical Name or Material Acetone-d6, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.