Product Code. 11399927
Click to view available options
Unit Size:
Pack of 10
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
11399927
10 Ea.
Pack of 10
11389927
2 Ea.
Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
11399927
Supplier
Thermo Scientific Alfa Aesar
Supplier No.
042260.KV
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
CAS
666-52-4
Molecular Formula
C3H6O
Molecular Weight (g/mol)
64.117
MDL Number
MFCD00044635
InChI Key
CSCPPACGZOOCGX-WFGJKAKNSA-N
Synonym
acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
PubChem CID
522220
ChEBI
CHEBI:78217
IUPAC Name
1,1,1,3,3,3-hexadeuteriopropan-2-one
SMILES
CC(=O)C
Specifications
Melting Point
-94°C
Density
0.872
Boiling Point
56°C
Flash Point
-17°C (1°F)
Packaging
Prescored ampoules, 0.75mL/ampoule
Refractive Index
1.3554
Quantity
10 Ea.
UN Number
UN1090
Specific Gravity
0.872
Solubility Information
Fully miscible in water.
Formula Weight
64.13
Percent Purity
100%
Physical Form
Liquid
Chemical Name or Material
Acetone-d6
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?