missing translation for 'onlineSavingsMsg'
Learn More

Acetone-d6, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D

Product Code. 10721921 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
7.5 mL
Packaging:
Ampule
Unit Size:
7.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10721921 7.5 mL Ampule 7.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10721921 Supplier Thermo Scientific Acros Supplier No. 320640075
  • ONLINE EXCLUSIVE PRICE
    97.35 €
    94.50 € / 7.5mL
    Save 2.85 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 666-52-4 | C3H6O | 64.117 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 0.8700g/mL
Deuterium (D) 99.4% min.
Flash Point −20°C
Packaging Ampule
Water 0.3% max. (Coulometric)
Quantity 7.5 mL
CAS 666-52-4
Melting Point -93.0°C
Boiling Point 55.0°C
Molecular Formula C3H6O
Linear Formula CD3COCD3
MDL Number MFCD00044635
Beilstein 01, III, 2741
Synonym acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
InChI Key CSCPPACGZOOCGX-WFGJKAKNSA-N
SMILES CC(=O)C
IUPAC Name 1,1,1,3,3,3-hexadeuteriopropan-2-one
Molecular Weight (g/mol) 64.117
PubChem CID 522220
ChEBI CHEBI:78217
Formula Weight 64.13
Grade NMR
Chemical Name or Material Acetone-d6, 99.5 atom % D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone-d6
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.