missing translation for 'onlineSavingsMsg'
Learn More

Acetone-d6, for NMR, packaged in 0.75ml ampoules, 100.0 atom % D

Product Code. 10339460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
7.5 mL
Packaging:
Ampule
Unit Size:
7.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10339460 7.5 mL Ampule 7.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10339460 Supplier Thermo Scientific Acros Supplier No. 320610075
  • ONLINE EXCLUSIVE PRICE
    115.00 €
    111.00 € / 7.5mL
    Save 4.00 €
    3% Off
Ver todas las promociones
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: es posible que la compra de este producto esté sujeta a restricciones y necesitemos solicitarle documentación adicional.

CAS: 666-52-4 | C3H6O | 64.117 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Especificaciones

Physical Form Liquid
Color Colorless
Density 0.8700g/mL
Deuterium (D) 99.95% min.
Flash Point −20°C
Packaging Ampule
Water 0.02% max. (Coulometric)
Quantity 7.5 mL
CAS 666-52-4
Melting Point -93.0°C
Boiling Point 55.0°C
Molecular Formula C3H6O
Linear Formula CD3COCD3
MDL Number MFCD00044635
Beilstein 01, III, 2741
Synonym acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
InChI Key CSCPPACGZOOCGX-WFGJKAKNSA-N
SMILES CC(=O)C
IUPAC Name 1,1,1,3,3,3-hexadeuteriopropan-2-one
Molecular Weight (g/mol) 64.117
PubChem CID 522220
ChEBI CHEBI:78217
Formula Weight 64.13
Grade NMR
Chemical Name or Material Acetone-d6, 100.0 atom % D
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • Acetone-d6
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Otros peligros
  • MIXTURE LIST-Contain: Acetone

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.