missing translation for 'onlineSavingsMsg'
Learn More

Acetone-d6 (reagent grade), CDN

Product Code. 30318452
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
30318452 50 g 50g
30309989 10 g 10g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30318452

Brand: CDN CDND000250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetone-D6
CAS 666-52-4
Molecular Formula C3 2H6 O
Quantity 50 g
InChI Formula InChI=1 S/C3H6O/c1-3(2)4/h1-2H3/i1D3,2D3
SMILES [2 H]C([2 H])([2 H])C(=O)C([2 H])([2 H])[2 H]
IUPAC Name 1,1,1,3,3,3-hexadeuteriopropan-2-one
Molecular Weight (g/mol) 64.12
Formula Weight 64.0795 g/mol
Percent Purity 99 atom % D, min 99% Chemical Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.