missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Laboratory Reagent, ≥99.5%, Solstice

Product Code. 15651640 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
25 L
4 x 4 L
4 x 5 L
6 x 1 L
Packaging:
Glass Bottle
Poly Bottle
Stainless Steel Can
Unit Size:
1 L
10 L
20 L
25 L
4 L
Pack of 4
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15651640 1 L Glass Bottle 1 L
15688320 4 L Glass Bottle 4 L
15611650 6 x 1 L Glass Bottle Pack of 6
15631640 10 L Stainless Steel Can 10 L
15691640 4 x 4 L Glass Bottle Pack of 4
15601650 4 x 5 L Poly Bottle Pack of 4
15671640 20 L Stainless Steel Can 20 L
15681640 25 L Stainless Steel Can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15651640 Supplier Fluka Supplier No. 1799731L
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Laboratory Reagent,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94°C
Color Colorless
Density 0.791g/mL at 25°C
Quantity 1 L
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.359(lit.)
Linear Formula CH3COCH3
Vapor Pressure 184mmHg (20°C)
UN Number UN1090
Beilstein 635680
Boiling Point 56°C
Formula Weight 58.08g/mol
Percent Purity ≥99.5%
Grade Laboratory Reagent
Chemical Name or Material Acetone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.