missing translation for 'onlineSavingsMsg'
Learn More

Acetone oxime, 98%

Product Code. 11409732
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11409732 250 g 250g
11499722 50 g 50g
11489722 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409732 Supplier Thermo Scientific Alfa Aesar Supplier No. A10802.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 127-06-0 | C3H7NO | 73.10 g/mol

It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.

Solubility
Soluble in water (330 g/L at 20°C).

Notes
Store away from oxidizing agents. Protect against electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-06-0
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00002118
InChI Key PXAJQJMDEXJWFB-UHFFFAOYSA-N
Synonym acetone oxime, acetoxime, propan-2-one oxime, 2-propanone, oxime, 2-propanone oxime, acetonoxime, acetone, oxime, beta-isonitrosopropane, acetoneoxime, ccris 5
PubChem CID 67180
ChEBI CHEBI:15349
IUPAC Name N-propan-2-ylidenehydroxylamine
SMILES CC(C)=NO

Specifications

Melting Point 60°C to 63°C
Density 0.901
Boiling Point 133°C to 135°C
Flash Point 60°C (140°F)
Quantity 250 g
UN Number UN1325
Beilstein 1560146
Merck Index 14,75
Solubility Information Soluble in water (330g/L at 20°C).
Formula Weight 73.1
Percent Purity 98%
Chemical Name or Material Acetone oxime
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.