missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Puriss., meets analytical spec. of BP, NF, Ph. Eur., ≥99% (GC), Solstice
BIOPHARMA

Product Code. 15632550
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Glass Bottle
Plastic Bottle
Stainless Steel Can
Unit Size:
1 L
10 L
2.5 L
25 L
5 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15632550 25 L Stainless Steel Can 25 L
15692540 1 L Plastic Bottle 1 L
15612550 2.5 L Glass Bottle 2.5 L
15602550 2.5 L Plastic Bottle 2.5 L
15672550 5 L Plastic Bottle 5 L
15672540 10 L Stainless Steel Can 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15632550 Supplier Honeywell-Riedel-de Haen Supplier No. 2420125L
  • ONLINE EXCLUSIVE PRICE
    770.00 €
    630.75 € / 25 L
    Save 139.25 €
    18% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Density 0.791g/mL at 25°C, 0.790 to 0.792g/mL at 20°C
Quantity 25 L
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Packaging Stainless Steel Can
Linear Formula CH3COCH3
Vapor Pressure 184mmHg (20°C)
UN Number 1090
Beilstein 635680
Formula Weight 58.08g/mol
Percent Purity ≥99% (GC)
Grade puriss.
Chemical Name or Material Acetone, Meets analytical specification of Ph. Eur.BP, NF
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.