missing translation for 'onlineSavingsMsg'
Learn More

Acetone, puriss. p.a., ACS Reagent, Reag. Ph. Eur. reag. ISO, ≥99.5% (GC), Solstice

Product Code. 15613680
Change view
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
Pack of 4
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15613680 6 x 1 L Poly Bottle Pack of 6
15603680 4 x 2.5 L Glass Bottle Pack of 4
15693670 4 x 2.5 L Poly Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15613680 Supplier Honeywell-Riedel-de Haen Supplier No. 322016X1L
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94°C
Appearance Colorless liquid
Density 0.791g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Packaging Poly Bottle
Refractive Index n20/D 1.359
Residue after Evaporation 0 to 0.001%
Titratable Acid ≤0.0015% (as CH3COOH)
Titratable Base ≤0.0008% (as NH3)
Linear Formula CH3COCH3
Vapor Pressure 184mmHg (20°C)
UN Number UN1090
Beilstein 635680
Boiling Point 56°C (760 mmHg)
Formula Weight 58.08g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Acetone, Reag. Ph. Eur., reag. ISO
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.