missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile with 0.1% formic acid, ≥99.5% (GC), Honeywell™ Riedel-de-Haën™ CHROMASOLV™

Product Code. 15614330
Change view
Click to view available options
Quantity:
1 L
4 x 4 L
Unit Size:
1 L
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15614330 1 L 1 L
15634330 4 x 4 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15614330 Supplier Honeywell Riedel-de Haën Supplier No. 346681L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

MDL Number MFCD00001878

Specifications

Quantity 1 L
Flash Point 2°C (35.6°F)
UN Number UN1648
Percent Purity ≥99.5% (GC)
Chemical Name or Material Acetonitrile with 0.1% formic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.