missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile with 0.1% Trifluoroacetic Acid (v/v), Certified HPLC for Gradient Analysis
Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Unit Size:
2.5 L
Specifications
Specifications
| CAS | 75-05-8 |
| Quantity | 2.5 L |
| Packaging | Amber Glass Bottle |
| Molecular Formula | C2HF3O2 |
| MDL Number | MFCD00004169 |
| Synonym | trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova |
| InChI Key | DTQVDTLACAAQTR-UHFFFAOYSA-N |
| SMILES | OC(=O)C(F)(F)F |
| IUPAC Name | trifluoroacetic acid |
| Molecular Weight (g/mol) | 114.02 |
| Show More |
Safety and Handling
Product Identifier
- Acetonitrile with 0,1% TFA( v/v) for HPLC Gradient analysis
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304-IF INHALED:.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P240-Ground/bond container and receiving equipment.
- MIXTURE LIST-Contains : Acetonitrile, Trifluoroacetic acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction