missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile,Solstice Biosyn™
BIOPHARMA

Product Code. 16861240
Change view
Click to view available options
Packaging:
Glass Bottle
Quantity:
2.5 L
4 x 2.5 L
Unit Size:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
16861240 Glass Bottle 4 x 2.5 L 10 L
16851240 Glass Bottle 2.5 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16861240 Supplier Burdick & Jackson BioSyn Supplier No. BB0174X2.5L
  • ONLINE EXCLUSIVE PRICE
    109.00 €
    97.73 € / 10 L
    Save 11.27 €
    10% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biosynthesis and applications that require low water content. Meets UV specifications, > 99.9% purity.

  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 41.05
Synonym Methyl Cyanide, Cyanomethane

Specifications

Melting Point -46°C
Absorbance 0.05 AU (200 nm)
Color Colorless
CAS 75-05-8
Density 0.7857 g/cm3 (20°C)
Quantity 4 x 2.5 L
Explosion Limit 3% to 16% (V)
Flash Point 6°C
CAS Max % 100
Packaging Glass Bottle
Physical Form Liquid
Product Line BioSyn
Refractive Index 1.3434 - 1.3446 (20°C)
Vapor Pressure 97.3 hPa (20°C)
UN Number UN1648
Boiling Point 82°C (1.013 hPa)
Solubility Information Completely Soluble (in water)
Percent Purity ≥99.9%
Grade Low Water
Chemical Name or Material Acetonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.