missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, 80%, 20% Water with 0.1% Formic Acid, Optima LC/MS, Fisher Chemical

Product Code. 15431423
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431423 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15431423 Supplier Fisher Chemical Supplier No. LS122500
  • ONLINE EXCLUSIVE PRICE
    163.00 €
    136.00 € / 500mL
    Save 27.00 €
    16% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Reduced concentration of acetonitrile improves chromatographic performance, especially of early eluting, hydrophilic peptides.
  • Pre-mixed for ease-of-use.
  • Low metal ion content to ensure minimal metal contamination.
  • Ultra filtered to ensure low levels of particulate contamination.
  • Recommended solvent for use with Thermo Scientific EASY-nLC 1200 and Ultimate RSLCnano.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 7732-18-5, 64-18-6
Quantity 500 mL
Odor Aromatic
Physical Form Liquid
Specific Gravity 0.7810
Molecular Formula C2H3N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.053
PubChem CID 6342
ChEBI CHEBI:38472
Chemical Name or Material Acetonitile with formic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 80% Acetonitrile, 20% Water with 0.1% Formic Acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contain: Acetonitrile, Formicacid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.