missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, 80%, 20% Water with 0.1% Formic Acid, Thermo Scientific

Product Code. 30280954 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280954 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280954 Supplier Thermo Scientific Supplier No. TL01220500
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 28-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Reduced concentration of acetonitrile improves chromatographic performance, especially of early eluting, hydrophilic peptides.
  • Pre-mixed for ease-of-use.
  • Low metal ion content to ensure minimal metal contamination.
  • Ultra filtered to ensure low levels of particulate contamination.
  • Recommended solvent for use with Thermo Scientific EASY-nLC 1200 and Ultimate RSLCnano.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7732-18-5, 64-18-6, 75-05-8
Quantity 500 mL
Packaging Glass Bottle
Grade LC-MS
Chemical Name or Material Acetonitrile, 80%, 20% Water with 0.1% Formic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.