missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™

Product Code. 15695050
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
10 L
2.5 L
20 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15695050 1 L Glass Bottle 1 L
15625060 500 mL Glass Bottle 500mL
15605060 2.5 L Glass Bottle 2.5 L
15685050 10 L Stainless Steel Can 10 L
15615060 20 L Stainless Steel Can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15695050 Supplier Honeywell-Riedel-de Haen Supplier No. 3604571L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C
Density 0.786g/mL at 25°C
Quantity 1 L
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Infrared Spectrum Conforms to structure
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0 to 0.005%
Titratable Acid ≤8μEQ/g
Titratable Base ≤0.6μEQ/g
Linear Formula CH3CN
Vapor Pressure 72.8mmHg (20°C)
UN Number UN1648
Beilstein 741857
Boiling Point 81°C
Formula Weight 41.05g/mol
Percent Purity ≥99.5%
Chemical Name or Material Acetonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.