missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, ChromAR™, Macron Fine Chemicals™

Product Code. 15315051
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15315051 2.5 L Narrow Mouth Amber Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15315051 Supplier Macron Fine Chemicals Supplier No. 285625

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Stored in an amber, narrow mouth glass bottle for protection from light. Macron Fine Chemicals™ ChromAR™ Acetonitrile meets ACS specifications and is suitable for liquid chromatography and UV-spectrophotometry.

Specifications

Melting Point −46°C
Absorbance Max. 1.00 at 190nm, Max. 0.05 at 200-210nm, Max. 0.02 at 220nm, Max. 0.01 at 230nm, Max. 0.005 at 254-400nm
CAS 75-05-8
Color Clear, colorless
Appearance Passes Test
Density 0.79g/mL at 20°C
Quantity 2.5 L
Explosion Limit Lower: 4.4% Vol, Upper: 16% Vol
Flash Point 2.00°C
Ignition Point 524°C
Odor Ether-like
Packaging Narrow Mouth Amber Glass Bottle
Physical Form Liquid
Residue after Evaporation Max. 0.0002%
Titratable Acid Max. 8μeq/g
Molecular Formula CH3CN
Titratable Base Max. 0.6μeq/g
Vapor Pressure 9.73 kPa at 20°C
Water Max. 0.02%, by Karl Fischer titration
Boiling Point 81°C
Molecular Weight (g/mol) 41.05
Percent Purity ≥99.8%
Grade ChromAR
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.