missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, CHROMASOLV™, ≥99.9%, for LC-MS, Honeywell Riedel-de Haën™

Product Code. 15624750
Change view
Click to view available options
Quantity:
4 x 4 L
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15624750 4 x 4 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15624750 Supplier Honeywell-Chromasolv Supplier No. 349674X4L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 75-05-8
Color Colorless
Density 0.786g/mL at 25°C
Quantity 4 x 4 L
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Molecular Formula C2H3N
Linear Formula CH3CN
MDL Number MFCD00001878
Vapor Pressure 72.8 mmHg (20°C)
UN Number UN1648
Beilstein 741857
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.053
PubChem CID 6342
ChEBI CHEBI:38472
Formula Weight 41.05g/mol
Percent Purity ≥99.9%
Grade LC-MS CHROMASOLV
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.