missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile-d{3}, 100% (Isotopic)

Product Code. 11329947
Change view
Click to view available options
Quantity:
10 Ea.
2 Ea.
Unit Size:
Pack of 10
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11329947 10 Ea. Pack of 10
11319947 2 Ea. Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11329947 Supplier Thermo Scientific Alfa Aesar Supplier No. 042266.KV
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 2206-26-0 | C2H3N | 44.07 g/mol

Isotope labeled Acetonitrile is used as a polar aprotic solvent in organic synthesis. Acetonitrile-d3 is used as IR and NMR solvent. Probe for zeolite acidity. Cartridge eluent in HPLC- SPE (solid-phase extraction)-NMR technique.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-26-0
Molecular Formula C2H3N
Molecular Weight (g/mol) 44.07
MDL Number MFCD00001881
InChI Key WEVYAHXRMPXWCK-FIBGUPNXSA-N
Synonym acetonitrile-d3, 2h3 acetonitrile, cd3cn, acetonitrile-2,2,2-d3, methyl-d3 cyanide, trideuteroacetonitrile, acetonitrile-d3, 99.8 atom % d, acetonitrile-d3, ≥99.8 atom % d, acetonitrile-d isotopic 5g
PubChem CID 123151
IUPAC Name 2,2,2-trideuterioacetonitrile
SMILES [2H]C([2H])([2H])C#N

Specifications

Physical Form Liquid
Color Colorless
Density 0.844 g/mL
Flash Point 5°C (42°F)
Odor Pungent
Sensitivity Hygroscopic
Quantity 10 Ea.
Boiling Point 81°C
UN Number UN1648
Merck Index 14,70
Solubility Information Fully miscible in water.
Formula Weight 44.08
Percent Purity 100%
Chemical Name or Material Acetonitrile-d3, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.