missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile-d3, for NMR, 100 atom % D, packaged in 0.75 ml ampoules

Product Code. 16398206 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
7.5 mL
Unit Size:
7.50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16398206 7.5 mL 7.50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16398206 Supplier Thermo Scientific Acros Supplier No. 464170075
  • ONLINE EXCLUSIVE PRICE
    206.00 €
    200.00 € / 7.5mL
    Save 6.00 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2206-26-0 | C2H3N | 44.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Density 0.8440 g/mL
Flash Point 2°C
Refractive Index 1.3430 to 1.3450
Specific Gravity 0.844
Viscosity 0.39 mPa.s (20°C)
Quantity 7.5 mL
CAS 2206-26-0
Melting Point -46.0°C
Boiling Point 79.0°C to 81.0°C
Molecular Formula C2H3N
Linear Formula CD3CN
MDL Number MFCD00001881
Solubility Information Solubility aq. soln.: soluble.
InChI Key WEVYAHXRMPXWCK-FIBGUPNXSA-N
SMILES [2H]C([2H])([2H])C#N
IUPAC Name (²H₃)acetonitrile
Molecular Weight (g/mol) 44.07
Formula Weight 44.07
Grade NMR
Chemical Name or Material Acetonitrile-d3
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.