missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile-d3, for NMR, 99.5 atom % D

Product Code. 16368196 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16368196 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16368196 Supplier Thermo Scientific Acros Supplier No. 389860100
  • ONLINE EXCLUSIVE PRICE
    69.75 €
    67.70 € / 10mL
    Save 2.05 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2206-26-0 | C2H3N | 44.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Density 0.8440 g/mL
Flash Point 2°C
Refractive Index 1.3430 to 1.3450
Specific Gravity 0.844
Viscosity 0.39 mPa.s (20°C)
Quantity 10 mL
CAS 2206-26-0
Melting Point -46.0°C
Boiling Point 79.0°C to 81.0°C
Molecular Formula C2H3N
Linear Formula CD3CN
MDL Number MFCD00001881
Solubility Information Solubility aq. soln.: soluble.
InChI Key WEVYAHXRMPXWCK-FIBGUPNXSA-N
SMILES [2H]C([2H])([2H])C#N
IUPAC Name (²H₃)acetonitrile
Molecular Weight (g/mol) 44.07
Formula Weight 44.07
Grade NMR
Chemical Name or Material Acetonitrile-d3
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile-d3, for NMR, 99.5 atom % D
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Acetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.