missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile-d3, for NMR, 99.9 atom% D

Product Code. 10284791 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Glass Bottle
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10284791 10 mL Glass Bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10284791 Supplier Thermo Scientific Acros Supplier No. 351390100
  • ONLINE EXCLUSIVE PRICE
    86.15 €
    83.60 € / 10mL
    Save 2.55 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 2206-26-0 | C2H3N | 44.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 0.8440g/mL
Deuterium (D) 99.85% min.
Flash Point 2°C
Packaging Glass Bottle
Refractive Index 1.3430 to 1.3450
Viscosity 0.39 mPa.s (20°C)
Water 0.03% max. (Coulometric)
Quantity 10 mL
CAS 2206-26-0
Melting Point -46.0°C
Boiling Point 79.0°C to 81.0°C
Molecular Formula C2H3N
Linear Formula CD3CN
MDL Number MFCD00001881
Beilstein 02, IV, 428
Synonym acetonitrile-d3, 2h3 acetonitrile, cd3cn, acetonitrile-2,2,2-d3, methyl-d3 cyanide, trideuteroacetonitrile, acetonitrile-d3, 99.8 atom % d, acetonitrile-d3, ≥99.8 atom % d, acetonitrile-d isotopic 5g
InChI Key WEVYAHXRMPXWCK-FIBGUPNXSA-N
SMILES [2H]C([2H])([2H])C#N
IUPAC Name 2,2,2-trideuterioacetonitrile
Molecular Weight (g/mol) 44.07
PubChem CID 123151
Formula Weight 44.07
Grade NMR
Chemical Name or Material Acetonitrile-d3, 99.9 atom% D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile-d3
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.