missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile-d3, for NMR, 99 atom % D

Product Code. p-3734290 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
50 mL
250 mL
Packaging:
Glass Bottle
Unit Size:
10mL
250mL
25mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10468730 10 mL Glass Bottle 10mL
10793161 25 mL Glass Bottle 25mL
10793351 50 mL Glass Bottle 50mL
10297443 250 mL Glass Bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10468730 Supplier Thermo Scientific Acros Supplier No. 166230100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Acetonitrile-d3, C2H3N, CAS Number-2206-26-0, 2h3 acetonitrile, acetonitrile-2,2,2-d3, acetonitrile-d3, acetonitrile-d3, >-99.8 atom % d, acetonitrile-d3, 99.8 atom % d, acetonitrile-d isotopic 5g, cd3cn, methyl-d3 cyanide, trideuteroacetonitrile, 10mL, 02, IV, 428, 79.0 deg.C to 81.0 deg.C | CAS: 2206-26-0 | C2H3N | 44.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 2206-26-0
Molecular Formula C2H3N
Molecular Weight (g/mol) 44.07
MDL Number MFCD00001881
InChI Key WEVYAHXRMPXWCK-FIBGUPNXSA-N
PubChem CID 123151
SMILES [2H]C([2H])([2H])C#N

Specifica

Physical Form Liquid
Color Colorless
Density 0.8440g/mL
Deuterium (D) 98.5% min.
Flash Point 2°C
Packaging Glass Bottle
Refractive Index 1.3430 to 1.3450
Viscosity 0.39 mPa.s (20°C)
Water 0.1% max. (Coulometric)
Quantity 10 mL
Melting Point -46.0°C
Boiling Point 79.0°C to 81.0°C
Linear Formula CD3CN
Beilstein 02, IV, 428
Synonym acetonitrile-d3, 2h3 acetonitrile, cd3cn, acetonitrile-2,2,2-d3, methyl-d3 cyanide, trideuteroacetonitrile, acetonitrile-d3, 99.8 atom % d, acetonitrile-d3, ≥99.8 atom % d, acetonitrile-d isotopic 5g
IUPAC Name 2,2,2-trideuterioacetonitrile
Formula Weight 44.07
Grade NMR
Chemical Name or Material Acetonitrile-d3, 99 atom%D
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • Acetonitrile-d3
Avvertenza
  • Danger
Categoria di pericolo
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Indicazioni di pericolo
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.