missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, for DNA Synthesis and Protein Sequencing, Fisher Chemical™

Product Code. 10510911 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 10510911

Brand: Fisher Chemical A/0628/17

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

C2H3N, CAS Number-75-05-8, 2.5L, 81 deg.C, Colorless, 41.05g/mol, -46 deg.C, 1878, Amber glass bottle, Liquid, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
MDL Number 1878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Color Colorless
Boiling Point 81°C
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 97mbar at 20°C
Viscosity 0.36cp at 20°C
Formula Weight 41.05g/mol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.