missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, far UV/gradient grade, 99.9+%

Product Code. 15416207
Change view
Click to view available options
Quantity:
1000 mL
2500 mL
Unit Size:
1000 L
2500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15416207 1000 mL 1000 L
15406207 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416207 Supplier GC Internal (Alfa Aesar) Supplier No. H33387.AU
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 75-05-8 | C{2}H{3}N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C{2}H{3}N
Synonym Methyl cyanide, Cyanomethane

Specifications

Melting Point −48°C to −45°C
Quantity 1000 mL
Boiling Point 80°C to 82°C
Percent Purity 99.9+%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.