missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, LC/MS grade, Thermo Scientific

Product Code. 30280939 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280939 1 L 1 L
30280938 500 mL 500mL
30280940 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30280939 Supplier Thermo Scientific Supplier No. T009551000
  • ONLINE EXCLUSIVE PRICE
    387.00 €
    283.00 € / 1 L
    Save 104.00 €
    26% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

Molecular Weight (g/mol) 41.05 g/mol

Specifications

Quantity 1 L
CAS 75-05-8
Packaging Glass Bottle
Boiling Point 81°C
Percent Purity 99.90%
Grade LC-MS
Chemical Name or Material Acetonitrile
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.