missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Ultra Gradient Grade, BAKER ANALYZED HPLC, J.T. Baker™

Product Code. p-7138045 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
5 L
Packaging:
EcoTainer Metal Safety Can
Glass
Unit Size:
2.5 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10231461 2.5 L Glass 2.5 L
10614471 5 L EcoTainer Metal Safety Can 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10231461 Supplier JT Baker Supplier No. 9017.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Get rapid, reproducible performance and separation results. Acetonitrile, Ultra Gradient Grade, BAKER ANALYZED HPLC, J.T. Baker™ is an application-optimized, function-tested HPLC reagent/solvent that maximizes throughput and ease-of-use without sacrificing efficiency or reproducibility.

BAKER ANALYZED HPLC Reagents are high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for research and quality control applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Recalibration or adjustments due to lot changes are minimized for optimum performance from sensitive instrumentation
  • Low backgrounds free of extraneous peaks
  • Low UV absorbance in critical ranges - assured through UV absorbance testing at a variety of points and through gradient elution testing
  • Fluorescence testing for trace impurities which would cause interference
  • Function-tested for: assay, water, minimal residue after evaporation and UV absorbance and fluorescence in critical ranges
  • Lot to lot consistency, with an actual Lot Analysis on the label
  • Innovative packaging options to assure solvent quality to the point-of-use

Specifications

Melting Point -46°C
Absorbance Max. 0.05 at 200nm, Max. 0.01 at 220nm, Max. 0.01 at 254-400nm
Color Colorless
CAS 75-05-8
Appearance Passes test
Boiling Point 81°C
Quantity 2.5 L
Explosion Limit 4.4 - 16 Vol %
Flash Point 5°C
Identification Passes test
Ignition Point 525°C
Odor Aromatic
Packaging Glass
Physical Form Liquid
Refractive Index 1.344 n20/d
Residue after Evaporation Max. 1 ppm
Molecular Formula C2H3N
Titratable Acid Max. 0.0008meq/g
Linear Formula CH3CN
Titratable Base Max. 0.0006meq/g
Viscosity 0.39 mPas at 20°C
Water Max. 0.01%
Synonym Ethanenitrile, Cyanomethane, Methyl cyanide
Solubility Information Fully miscible (in water)
Formula Weight 41.05g/mol
Percent Purity ≥99.9%
Grade Ultra Gradient HPLC Grade
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.