missing translation for 'onlineSavingsMsg'
Learn More

Acetonylacetone, 97%

Product Code. 11974811 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
250mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11974811 1 L Glass bottle 1 L
10050720 5 mL Glass bottle 5mL
10578090 100 mL Glass bottle 100mL
10407860 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11974811 Supplier Thermo Scientific Acros Supplier No. 102400010
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 110-13-4 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-13-4
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00008792
InChI Key OJVAMHKKJGICOG-UHFFFAOYSA-N
Synonym 2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane
PubChem CID 8035
ChEBI CHEBI:85014
IUPAC Name hexane-2,5-dione
SMILES CC(=O)CCC(=O)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -6.0°C
Color Yellow to Brown
Density 0.9730g/mL
Boiling Point 185.0°C to 193.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3COCH2CH2COCH3
Refractive Index 1.4250 to 1.4270
Quantity 1 L
Beilstein 01,788
Merck Index 15,7
Specific Gravity 0.973
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, ether, ethanol and, diethyl ether, miscible in organic solvents
Viscosity 1.43 mPa.s (25°C)
Formula Weight 114.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Acetonylacetone, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonylacetone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.