missing translation for 'onlineSavingsMsg'
Learn More

Acetophenazine dimaleate, MedChemExpress

Product Code. 16488706
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488706 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488706 Supplier Medchem Express Supplier No. HYB1262
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Acetophenazine dimaleate, a phenothiazine derivative, is an antipsychotic agent. Acetophenazine dimaleate primarily blocks dopamine D2 receptors in the brain. Acetophenazine dimaleate can be used for researching psychotic disorders such as schizophrenia and anxious depression.

Acetophenazine dimaleate, a phenothiazine derivative, is an antipsychotic agent. Acetophenazine dimaleate primarily blocks dopamine D2 receptors in the brain. Acetophenazine dimaleate can be used for researching psychotic disorders such as schizophrenia and anxious depression.

Specifications

Chemical Name or Material Acetophenazine dimaleate
CAS 5714-00-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H37N3O10S
Quantity 100 mg
Solubility Information DMSO : 125 mg/mL (194.19 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (12.94 mM; ultrasonic and warming and heat to 60°C)
SMILES CC(C(C=C1N2CCCN3CCN(CCO)CC3)=CC=C1SC4=C2C=CC=C4)=O.O=C(O)/C=C\C(O)=O.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 643.7
Formula Weight 643.7
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.