missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetophenone-2',3',4',5',6'-d5, CDN
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 1-(2,3,4,5,6-pentadeuteriophenyl)ethanone |
| CAS | 28077-64-7 |
| Molecular Formula | C8 2H5 H3 O |
| Quantity | 5 g |
| InChI Formula | InChI=1 S/C8H8O/c1-7(9)8-5-3-2-4-6-8/h2-6 H,1H3/i2D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)C)c([2 H])c1[2 H] |
| IUPAC Name | 1-(2,3,4,5,6-pentadeuteriophenyl)ethanone |
| Molecular Weight (g/mol) | 125.179 |
| Formula Weight | 125.089 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction