missing translation for 'onlineSavingsMsg'
Learn More

Acetophenone, 99%

Product Code. 15453057
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453057 2500 mL 2500mL
15463057 500 mL 500mL
15443057 10,000 mL 10000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15453057 Supplier Thermo Scientific Alfa Aesar Supplier No. A12727.0F
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-86-2 | C8H8O | 120.151 g/mol

Acetophenone is used as a raw material in the synthesis of active pharmaceutical ingredients such as dextropropoxyphene and phenylpropanolamine. It is employed in organic synthesis and as a polymerization catalyst for olefins. It is involved in the preparation of styrene, 1-phenylethanol. It is also used as a active component of coatings and inks. It reacts with formaldehyde to prepare resins. Further, it is used in textile coatings, in perfumes for orange blossom odor. In addition to this, it is used as a solvent in plastics, adhesives and a flavoring agent in foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetophenone is used as a raw material in the synthesis of active pharmaceutical ingredients such as dextropropoxyphene and phenylpropanolamine. It is employed in organic synthesis and as a polymerization catalyst for olefins. It is involved in the preparation of styrene, 1-phenylethanol. It is also used as a active component of coatings and inks. It reacts with formaldehyde to prepare resins. Further, it is used in textile coatings, in perfumes for orange blossom odor. In addition to this, it is used as a solvent in plastics, adhesives and a flavoring agent in foods.

Solubility
Miscible with water, chloroform, ethanol, ether, benzene and acetone. Slightly miscible with concentrated sulfuric acid.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-86-2
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00008724
InChI Key KWOLFJPFCHCOCG-UHFFFAOYSA-N
Synonym acetophenone, methyl phenyl ketone, acetylbenzene, phenyl methyl ketone, ethanone, 1-phenyl, hypnone, benzoyl methide, acetophenon, 1-phenylethan-1-one, acetylbenzol
PubChem CID 7410
ChEBI CHEBI:27632
IUPAC Name 1-phenylethanone
SMILES CC(=O)C1=CC=CC=C1

Specifications

Melting Point 19°C to 20°C
Density 1.03
Boiling Point 201°C to 203°C
Flash Point 76°C (169°F)
Linear Formula C6H5COCH3
Refractive Index 1.5325
Quantity 2500 mL
Merck Index 14,73
Solubility Information Miscible with water,chloroform,ethanol,ether,benzene and acetone. Slightly miscible with concentrated sulfuric acid.
Formula Weight 120.15
Percent Purity 99%
Chemical Name or Material Acetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.