missing translation for 'onlineSavingsMsg'
Learn More

Acetophenone azine, 97%

Product Code. 15478819
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15478819 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15478819 Supplier Thermo Scientific Alfa Aesar Supplier No. L03924.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 729-43-1 | C16H16N2 | 236.318 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 729-43-1
Melting Point 120°C to 124°C
Molecular Formula C16H16N2
MDL Number MFCD00015011
Quantity 5 g
Beilstein 1912471
Synonym acetophenone azine, 1-phenyl-n-z-1-phenylethylideneamino ethanimine
InChI Key MOKMQSIJAHPSQX-SIJTWYJSSA-N
SMILES CC(=NN=C(C)C1=CC=CC=C1)C2=CC=CC=C2
IUPAC Name (E)-1-phenyl-N-[(Z)-1-phenylethylideneamino]ethanimine
Molecular Weight (g/mol) 236.318
PubChem CID 5914692
Formula Weight 236.32
Percent Purity 97%
Chemical Name or Material Acetophenone azine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.