missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Molecular Weight: 196.2 g/mol
- Purity: >98%
- Form: Powder
- CAS Number: 2478-38-8
- Key Features: Naturally occurring phenolic compound; inducer of Agrobacterium tumefaciens virulence genes; utilized in the synthesis of anticancer agents in vitro
- Storage: Store at ambient temperature
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% Acetosyringone |
| Content And Storage | Store at ambient temperature. |
| Format | Vial |
| Form | Powder |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Virulence Genes (Agrobacterium tumefaciens) |
| Formulation | 100% Acetosyringone |
| Quantity | 1 g |
| For Use With (Application) | Plant-pathogen recognition, Agrobacterium tumefaciens virulence gene induction, anticancer agent research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?