missing translation for 'onlineSavingsMsg'
Learn More

Acetoxyacetone, TRC

Product Code. 17948714
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17948714 10 g 10g
17918054 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17948714 Supplier Toronto Research Chemicals Supplier No. TRCA13303510G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-20-1
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
Synonym Acetoxypropanone, Acetylmethyl Acetate, Acetyloxyacetone, NSC 2298, NSC 7614, O-Acetylacetol, 1-(Acetyloxy)propan-2-one, 1-Acetoxy-2-propanone, 1-Acetoxyacetone, 1-Hydroxy-2-propanone Acetate, 2-Oxopropyl Acetate, Acetic Acid Acetonyl Ester, Acetol Acetate, Acetonyl Acetate;
IUPAC Name 2-oxopropyl acetate
SMILES CC(=O)COC(=O)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H8O3/c1-4(6)3-8-5(2)7/h3H2,1-2H3
Formula Weight 116.0473
Chemical Name or Material Acetoxyacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.