missing translation for 'onlineSavingsMsg'
Learn More

Acetyl(2-pyridinylmethyl)carbamic Acid 2-Methoxy-3-[[(octadecylamino)carbonyl]oxy]propyl Ester, TRC

Product Code. 30316223
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316223 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316223 Supplier Toronto Research Chemicals Supplier No. TRCA1879651MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetyl(2-pyridinylmethyl)carbamic Acid 2-Methoxy-3-(((octadecylamino)carbonyl)oxy)propyl Ester
CAS 100488-81-1
Molecular Formula C32H55N3O6
Quantity 1 mg
InChI Formula InChI=1S/C32H55N3O6/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-20-24-34-31(37)40-26-30(39-3)27-41-32(38)35(28(2)36)25-29-22-19-21-23-33-29/h19,21-23,30H,4-18,20,24-27H2,1-3H3,(H,34,37)
SMILES CCCCCCCCCCCCCCCCCCNC(=O)OCC(COC(=O)N(Cc1ccccn1)C(=O)C)OC
IUPAC Name [2-methoxy-3-(octadecylcarbamoyloxy)propyl] N-acetyl-N-(pyridin-2-ylmethyl)carbamate
Molecular Weight (g/mol) 577.8
Formula Weight 577.4091
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.