missing translation for 'onlineSavingsMsg'
Learn More

Acetyl chloride, 99+%

Product Code. 10772954 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
25mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10772954 2.5 L Glass bottle 2.5 L
10687352 25 mL Glass bottle 25mL
10308550 250 mL Glass bottle 250mL
10732384 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10772954 Supplier Thermo Scientific Acros Supplier No. 219470025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Acetyl chloride, 99+%, C2H3ClO, CAS Number-75-36-5, acetic acid chloride, acetic acid, chloride, acetic chloride, acetylchloride, ccris 4568, ch3cocl, ethanoyl chloride, hsdb 662, rcra waste number u006, unii-qd15rno45k, 250mL, 99% min. (GC), 02, 173, 51.0 deg.C, 100.0, 99.0, CHEBI:37580 | CAS: 75-36-5 | C2H3ClO | 78.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-36-5
Molecular Formula C2H3ClO
Molecular Weight (g/mol) 78.50
MDL Number MFCD00000719
InChI Key WETWJCDKMRHUPV-UHFFFAOYSA-N
Synonym ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662
PubChem CID 6367
ChEBI CHEBI:37580
IUPAC Name acetyl chloride
SMILES CC(Cl)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -112.0°C
Density 1.1000g/mL
Boiling Point 51.0°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCl
Refractive Index 1.3876 to 1.3896
Quantity 2.5 L
Beilstein 02, 173
Fieser 01,11; 04,5; 05,4; 07,3; 11,4
Merck Index 15, 80
Specific Gravity 1.1
Solubility Information Solubility in water: reacts. Other solubilities: miscible with acetone, benzenz, toluene,, chloroform, ether, petroleum ether, carbon dosulfi, de and glacial acetic acid, soluble in ccl4, reacts with alcohol
Formula Weight 78.5
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Acetyl chloride, p.a.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetyl chloride
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.