missing translation for 'onlineSavingsMsg'
Learn More

Acetyl chloride, 99+%

Product Code. 15421887
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421887 1 L 1 L
11399358 50 mL 50mL
11389358 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 15421887 Supplier Thermo Scientific Alfa Aesar Supplier No. 043262.K2
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 75-36-5 | C2H3ClO | 78.50 g/mol

Acetyl chloride acts as a reagent for the preparation of esters and amides of acetic acid. It is also useful an important reactant in Friedel-Crafts reactions as well as the introduction of an acetyl group. It serves as a starting material in the production of pharmaceutical, new plating complexing agent, acylation agent and synthetic organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-36-5
Molecular Formula C2H3ClO
Molecular Weight (g/mol) 78.50
MDL Number MFCD00000719
InChI Key WETWJCDKMRHUPV-UHFFFAOYSA-N
Synonym ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662
PubChem CID 6367
ChEBI CHEBI:37580
IUPAC Name acetyl chloride
SMILES CC(Cl)=O

Specifications

Melting Point -112°C
Color Colorless
Density 1.104
Boiling Point 50°C to 52°C
Flash Point 4°C (39°F)
Odor Pungent
Linear Formula CH3COCl
Refractive Index 1.389
Quantity 1 L
UN Number UN1717
Beilstein 605303
Sensitivity Moisture sensitive
Merck Index 14,85
Solubility Information Miscible with acetone,chloroform,glacial acetic acid,petroleum ether,ether and benzene.
Formula Weight 78.5
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Acetyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.